MilliporeSigma
  • Effect of erythrosine- and LED-mediated photodynamic therapy on buccal candidiasis infection of immunosuppressed mice and Candida albicans adherence to buccal epithelial cells.
Effect of erythrosine- and LED-mediated photodynamic therapy on buccal candidiasis infection of immunosuppressed mice and Candida albicans adherence to buccal epithelial cells.
Oral surgery, oral medicine, oral pathology and oral radiology (2012-06-26)
Anna Carolina Borges Pereira Costa, Vanessa Maria Campos Rasteiro, Emily Setsuko Halter da Silva Hashimoto, Cássia Fernandes Araújo, Cristiane Aparecida Pereira, Juliana Campos Junqueira, Antonio Olavo Cardoso Jorge
ABSTRACT
This study evaluated the effects of photodynamic therapy (PDT) on buccal candidiasis in mice and on the adherence of yeast to buccal epithelial cells (BECs) in vitro. A total of 56 immunosuppressed mice with buccal candidiasis were subjected to PDT, consisting of treatment with erythrosine (400 μmol/L) followed by exposure to a green LED (14.34 J cm(-2)). After treatment, the yeasts recovered from the mice were quantified (CFU/mL) and analyzed for the effects of PDT on their adherence to BECs. The data were analyzed using ANOVA, the Tukey test, Kruskal-Wallis test and Student t test. PDT significantly reduced the amount of yeast present in the lesions by 0.73 log(10) (P = .018) and reduced C. albicans adherence to BECs by 35% without damaging adjacent tissues (P = .045). Photodynamic therapy exhibited antifungal effects against C. albicans biofilms formed in vivo and reduced the capacity of C. albicans to adhere to BECs in vitro.
Sigma-Aldrich
Erythrosin B, certified by the Biological Stain Commission, Dye content ≥85 %
Sigma-Aldrich
Erythrosin extra bluish, for microscopy (Bact., Hist.), adsorption and fluorescent indicator
Sigma-Aldrich
Erythrosin extra bluish, certified by the Biological Stain Commission
Supelco
Erythrosin B, analytical standard