MilliporeSigma
  • Enzymatic transformation of the major ginsenoside Rb2 to minor compound Y and compound K by a ginsenoside-hydrolyzing β-glycosidase from Microbacterium esteraromaticum.
Enzymatic transformation of the major ginsenoside Rb2 to minor compound Y and compound K by a ginsenoside-hydrolyzing β-glycosidase from Microbacterium esteraromaticum.
Journal of industrial microbiology & biotechnology (2012-06-22)
Lin-Hu Quan, Yan Jin, Chao Wang, Jin-Woo Min, Yeon-Ju Kim, Deok-Chun Yang
ABSTRACT
The ginsenoside-hydrolyzing β-glycosidase (Bgp3) derived from Microbacterium esteraromaticum transformed the major ginsenoside Rb2 to more pharmacologically active minor ginsenosides including compounds Y and K. The bgp3 gene consists of 2,271 bp encoding 756 amino acids which have homology to the glycosyl hydrolase family 3 protein domain. Bgp3 is capable of hydrolyzing beta-glucose links and arabinose links. HPLC analysis of the time course of ginsenoside Rb2 hydrolysis by Bgp3 (0.1 mg enzyme ml(-1) in 20 mM sodium phosphate buffer at 40 °C and pH 7.0) showed that the glycosidase first hydrolyzed the inner glucose moiety attached to the C-3 position and then the arabinopyranose moiety attached to the C-20 position. Thus, Bgp3 hydrolyzed the ginsenoside Rb2 via the following pathway: Rb2 → compound Y → compound K.