MilliporeSigma
  • Vitamin E decreases extra-hepatic menaquinone-4 concentrations in rats fed menadione or phylloquinone.
Vitamin E decreases extra-hepatic menaquinone-4 concentrations in rats fed menadione or phylloquinone.
Molecular nutrition & food research (2012-06-19)
Sherry M Farley, Scott W Leonard, Edwin M Labut, Hannah F Raines, David J Card, Dominic J Harrington, Debbie J Mustacich, Maret G Traber
ABSTRACT
The mechanism for increased bleeding and decreased vitamin K status accompanying vitamin E supplementation is unknown. We hypothesized that elevated hepatic α-tocopherol (α-T) concentrations may stimulate vitamin K metabolism and excretion. Furthermore, α-T may interfere with the side chain removal of phylloquinone (PK) to form menadione (MN) as an intermediate for synthesis of tissue-specific menaquinone-4 (MK-4). In order to investigate these hypotheses, rats were fed phylloquinone (PK) or menadione (MN) containing diets (2 μmol/kg) for 2.5 weeks. From day 10, rats were given daily subcutaneous injections of either α-T (100 mg/kg) or vehicle and were sacrificed 24 h after the seventh injection. Irrespective of diet, α-T injections decreased MK-4 concentrations in brain, lung, kidney, and heart; and PK in lung. These decreases were not accompanied by increased excretion of urinary 5C- or 7C-aglycone vitamin K metabolites, however, the urinary α-T metabolite (α-CEHC) increased ≥ 100-fold. Moreover, α-T increases were accompanied by downregulation of hepatic cytochrome P450 expression and modified expression of tissue ATP-binding cassette transporters. Thus, in rats, high tissue α-T depleted tissue MK-4 without significantly increasing urinary vitamin K metabolite excretion. Changes in tissue MK-4 and PK levels may be a result of altered regulation of transporters.
Product Description
Supelco
Menaquinone (K2), analytical standard
Supelco
Phylloquinone (K1), analytical standard
Supelco
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vitamin K2
Sigma-Aldrich
Vitamin K1, viscous liquid
Phytomenadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers