MilliporeSigma
  • A new thermostable β-glucosidase mined from Dictyoglomus thermophilum: properties and performance in octyl glucoside synthesis at high temperatures.
A new thermostable β-glucosidase mined from Dictyoglomus thermophilum: properties and performance in octyl glucoside synthesis at high temperatures.
Bioresource technology (2012-06-19)
Zheng-Zheng Zou, Hui-Lei Yu, Chun-Xiu Li, Xin-Wen Zhou, Chihiro Hayashi, Jie Sun, Bao-Hong Liu, Tadeyuki Imanaka, Jian-He Xu
ABSTRACT
A new β-glucosidase (DtGH) representing 40% identity with an apple seed glycosidase (ASG) was cloned from Dictyoglomus thermophilum. DtGH showed extremely high thermostability in aqueous solution, with half-lives of 533, 44, and 5 h measured at 70, 80 and 90 °C, respectively. Therefore it was used for direct glycosylation of n-octanol at 70 °C instead of 50 °C as usually. As a result, the glucose based conversion was increased by 27%, but the time spent to reach equilibrium was decreased from 7 d to 3 d. This enzyme also exhibited excellent stability under the reaction environment, retaining 70-80% of its initial activity after 7 d of incubation at 70 °C in either 1.7 M glucose solution or octanol-aqueous (85:15, v/v) system. It could retain part of synthetic activity even in boiling water. Owing to the strong glucose-tolerance and extremely high thermostability, DtGH should be promising for various glucosides synthesis.
Sigma-Aldrich
Octyl-β-D-glucopyranoside 100 mM solution
Sigma-Aldrich
Octyl β-D-glucopyranoside solution, ≥95% (HPLC), 50 % (w/v) in H2O
Sigma-Aldrich
Octyl β-D-glucopyranoside, ≥98% (GC)
Sigma-Aldrich
Octyl β-D-glucopyranoside, BioXtra, ≥98% (GC)