MilliporeSigma
  • Effect of sulfate on anaerobic reduction of nitrobenzene with acetate or propionate as an electron donor.
Effect of sulfate on anaerobic reduction of nitrobenzene with acetate or propionate as an electron donor.
Water research (2012-06-19)
Jingang Huang, Yue Wen, Ning Ding, Yue Xu, Qi Zhou
ABSTRACT
Sulfate is frequently found in wastewaters that contain nitrobenzene. To reveal the effect of sulfate on the reductive transformation of nitrobenzene to aniline--with acetate or propionate as potential electron donors in anaerobic systems--an acetate series (R1-R5) and a propionate series (R6-R10) were set up. Each of these was comprised of five laboratory-scale sequence batch reactors. The two series were amended with the same amount of nitrobenzene and electron donor electron equivalents, whereas with increasing sulfate concentrations. Results indicated that the presence of sulfate could depress nitrobenzene reduction. Such depression is linked to the inhibition of nitroreductase activity and/or the shift of electron flow. In the acetate series, although sulfate did not strongly compete with nitrobenzene for electron donors, noncompetitive inhibition of specific nitrobenzene reduction rates by sulfate was observed, with an inhibition constant of 0.40 mM. Propionate, which can produce intermediate H₂ as preferred reducing equivalent, is a more effective primary electron donor for nitrobenzene reduction as compared to acetate. In the propionate series, sulfate was found to be a preferential electron acceptor as compared to nitrobenzene, resulting in a quick depletion of propionate and then a likely termination of H₂-releasing under higher sulfate concentrations (R9 and R10). In such a situation, nitrobenzene reduction slowed down, occurring two-stage zero-order kinetics.
Product Description
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%