MilliporeSigma
  • Iron phosphate microflowers as peroxidase mimic and superoxide dismutase mimic for biocatalysis and biosensing.
Iron phosphate microflowers as peroxidase mimic and superoxide dismutase mimic for biocatalysis and biosensing.
Chemical communications (Cambridge, England) (2012-06-16)
Wei Wang, Xiangpeng Jiang, Kezheng Chen
ABSTRACT
Novel iron phosphates microflowers which show SOD-like and peroxidase-like mimic activities were prepared, suggesting potential applications as a biocatalyst and a biosensor for H(2)O(2).
Sigma-Aldrich
Iron(III) phosphate dihydrate, Fe 29 %