MilliporeSigma
  • Nanohybrids of silver particles immobilized on silicate platelet for infected wound healing.
Nanohybrids of silver particles immobilized on silicate platelet for infected wound healing.
PloS one (2012-06-14)
Chia-Yu Chu, Fu-Chuo Peng, Ying-Fang Chiu, Hsing-Chuan Lee, Chien-Wen Chen, Jiun-Chiou Wei, Jiang-Jen Lin
ABSTRACT
Silver nanoparticles supported on nanoscale silicate platelets (AgNP/NSP) possess interesting properties, including a large surface area and high biocide effectiveness. The nanohybrid of AgNP/NSP at a weight ratio 7/93 contains 5-nm Ag particles supported on the surface of platelets with dimensions of approximately 80×80×1 nm(3). The nanohybrid expresses a trend of lower cytotoxicity at the concentration of 8.75 ppm Ag and low genotoxicity. Compared with conventional silver ions and the organically dispersed AgNPs, the nanohybrid promotes wound healing. We investigated overall wound healing by using acute burn and excision wound healing models. Tests on both infected wound models of mice were compared among the AgNP/NSP, polymer-dispersed AgNPs, the commercially available Aquacel, and silver sulfadiazine. The AgNP/NSP nanohybrid was superior for wound appearance, but had similar wound healing rates, vascular endothelial growth factor (VEGF)-A levels and transforming growth factor (TGF)-β1 expressions to Aquacel and silver sulfadiazine.
Sigma-Aldrich
Silver(I) sulfadiazine, 98%