MilliporeSigma
On-demand separation of oil-water mixtures.
Advanced materials (Deerfield Beach, Fla.) (2012-06-13)
Gibum Kwon, Arun K Kota, Yongxin Li, Ameya Sohani, Joseph M Mabry, Anish Tuteja
ABSTRACT
In this work, the first-ever membrane-based single unit operation that enables gravity driven, on-demand separation of various oil-water mixtures is developed. Using this methodology, the on-demand separation of free oil and water, oil-in-water emulsions, and water-in-oil emulsions is demonstrated, with ≥99.9% separation efficiency. A scaled-up apparatus to separate larger quantities (several liters) of oil-water emulsions is also developed.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Supelco
Hexadecane, analytical standard
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%