MilliporeSigma
  • Chemoenzymatic synthesis of the alarm pheromone (+)-verbenone from geranyl diphosphate.
Chemoenzymatic synthesis of the alarm pheromone (+)-verbenone from geranyl diphosphate.
Chemical communications (Cambridge, England) (2012-06-12)
Zulfa Yoosuf-Aly, Juan A Faraldos, David J Miller, Rudolf K Allemann
ABSTRACT
The enzyme-guided asymmetric synthesis of (+)-verbenone from geranyl diphosphate in a simple two-step, one pot transformation highlights the potential of chemoenzymatic procedures for the generation of high-value terpenoids.
Sigma-Aldrich
(1S)-(−)-Verbenone, 94%
Sigma-Aldrich
(1S)-(−)-Verbenone, ≥93%
Supelco
(1S)-(−)-Verbenone, analytical standard