MilliporeSigma
  • [The effect of ionizing radiation with low dose rate on the state of electron transfer chain of enterocyte mitochondria of rat small intestine].
[The effect of ionizing radiation with low dose rate on the state of electron transfer chain of enterocyte mitochondria of rat small intestine].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2012-06-12)
L V Grubskaia, V M Voĭtsitskiĭ, S V Khizhniak
ABSTRACT
The influence of ionizing radiation with low absorbed dose rate (55 mGy x min(-1)) in 1, 12 and 24 hours after irradiation in doses of 0.1; 0.5 and 1.0 Gy on functional state of the electron transfer chain of the rat small intestine mitochondria was investigated by assessment of the oxygen consumption rate. The uncoupling of oxidation and phosphorylation, a decrease of phosphorylation rate and inhibition of ATP hydrolysis reactions were established in mitochondria in dependence on the irradiation dose and time interval after irradiation. The functional peculiarities of the oxidation-phosphorylation coupling sites of the mitochondrial electron transfer chain were detected.
Product Description
Supelco
2,4-Dinitrophenol solution, reference material, 5000 μg/mL in methanol
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard