MilliporeSigma
Assessment of gelatinases (MMP-2 and MMP-9) by gelatin zymography.
Methods in molecular biology (Clifton, N.J.) (2012-06-08)
Marta Toth, Anjum Sohail, Rafael Fridman
ABSTRACT
Gelatin zymography is a simple yet powerful method to detect proteolytic enzymes capable of degrading gelatin from various biological sources. It is particularly useful for the assessment of two key members of the matrix metalloproteinase family, MMP-2 (gelatinase A) and MMP-9 (gelatinase B), due to their potent gelatin-degrading activity. This polyacrylamide gel electrophoresis-based method can provide a reliable assessment of the type of gelatinase, relative amount, and activation status (latent, compared with active enzyme forms) in cultured cells, tissues, and biological fluids. The method can be used to investigate factors that regulate gelatinase expression and modulate zymogen activation in experimental systems. The system provides information on the pattern of gelatinase expression and activation in human cancer tissues and how this relates to cancer progression. Interpretation of the data obtained in gelatin zymography requires a thorough understanding of the principles and pitfalls of the technique; this is particularly important when evaluating enzyme levels and the presence of active gelatinase species. If properly used, gelatin zymography is an excellent tool for the study of gelatinases in biological systems.
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Triton X-114, laboratory grade
Supelco
IGEPAL® CA-630
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
Triton X-405 solution, 70% in H2O