MilliporeSigma
  • Effects of chemical modification of sphingomyelin ammonium group on formation of liquid-ordered phase.
Effects of chemical modification of sphingomyelin ammonium group on formation of liquid-ordered phase.
Bioorganic & medicinal chemistry (2012-06-08)
Sarah A Goretta, Masanao Kinoshita, Shoko Mori, Hiroshi Tsuchikawa, Nobuaki Matsumori, Michio Murata
ABSTRACT
Sphingomyelin (SM) and cholesterol form microdomains called lipid rafts in cellular membranes. To develop a versatile fluorescent lipid probe, chemical modifications to both the hydrophobic and hydrophilic portions of SM are essential. Few reports describing SM probes with a fluorophore at the polar head group have been published. This study examined the effect of substitution on an ammonium moiety of SM on the membrane properties of SM. Two SM analogs with small propargyl and allyl groups on the quaternary nitrogen atom were synthesized and subjected to analysis using differential scanning calorimetry, fluorescent anisotropy, detergent solubilization, surface pressure, and density measurements. Results demonstrated that the two SM analogs retained the membrane properties of SM, including formation of an ordered phase and the ability to interact with cholesterol. A dansyl-substituted SM was prepared for fluorescent measurements. Dansyl-SM showed less of a propensity to form microdomains. These findings imply the potential application of N-substituted SMs as a raft-specific molecular probe.
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)