MilliporeSigma
  • Chiral phosphoric acid catalyzed inverse-electron-demand aza-Diels-Alder reaction of isoeugenol derivatives.
Chiral phosphoric acid catalyzed inverse-electron-demand aza-Diels-Alder reaction of isoeugenol derivatives.
Organic letters (2012-06-08)
Long He, Mathieu Bekkaye, Pascal Retailleau, Géraldine Masson
ABSTRACT
Highly enantio- and diastereoselective three-component inverse electron-demand aza-Diels-Alder reaction of aldehydes, anilines, and isoeugenol derivatives catalyzed by a chiral phosphoric acid catalyst are reported. A wide variety of 2,3,4-trisubstituted tetrahydroquinolines containing an aryl group at the 4-position were obtained in a one-pot process with good to high yields and excellent stereoselectivities (>95:5 dr and up to >99% ee).
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Supelco
Isoeugenol, analytical standard