MilliporeSigma
Microbial transformations of 7-hydroxyflavanone.
TheScientificWorldJournal (2012-06-02)
Edyta Kostrzewa-Susłow, Tomasz Janeczko
ABSTRACT
Microbial transformations of racemic 7-hydroxyflavanone using strains of genus Aspergillus (A. niger KB, A. niger 13/5, A. ochraceus 456) and the species Penicillium chermesinum 113 were studied. The products of O-methylation, O-methylation along with hydroxylation at C-3' and C-4', reduction of the carbonyl group, reduction of the carbonyl group along with hydroxylation at C-5, and dehydrogenation of C-2 and C-3 were obtained. Most of the products (with the exception of the O-methylation one) have stronger antioxidant properties than the initial substrate.
Sigma-Aldrich
7-Hydroxyflavanone, 98%