MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Farnesoid X receptor targeting to treat nonalcoholic steatohepatitis.
Drug discovery today (2012-06-02)
Luciano Adorini, Mark Pruzanski, David Shapiro
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is a highly prevalent chronic liver condition evolving in a proportion of patients into nonalcoholic steatohepatitis (NASH), an aggressive form of NAFLD associated with increased cardiovascular mortality and significant risk of progressive liver disease, including fibrosis, cirrhosis and hepatocellular carcinoma. At present, no specific therapies for NASH exist. In this review, we examine the evidence supporting activation of the farnesoid X receptor (FXR), a nuclear hormone receptor regulated by bile acids (BAs), for the treatment of NASH. We also discuss the potential of the semi-synthetic BA derivative obeticholic acid (OCA), a first-in-class FXR agonist, as a safe and effective drug to address this significant unmet medical need.
Product Description
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chenodeoxycholic acid