MilliporeSigma
  • Acetylphosphonate as a surrogate of acetate or acetamide in organocatalyzed enantioselective aldol reactions.
Acetylphosphonate as a surrogate of acetate or acetamide in organocatalyzed enantioselective aldol reactions.
Organic letters (2012-06-02)
Jie Guang, Qunsheng Guo, John Cong-Gui Zhao
ABSTRACT
Highly enantioselective aldol reactions of acetylphosphonates and activated carbonyl compounds was realized with cinchona alkaloid derived catalysts, in which the acetylphosphonate was directly used as an enolate precursor for the first time. The aldol product obtained was converted in situ to its corresponding ester or amide through methanolysis or aminolysis. The overall process may be viewed as formal highly enantioselective acetate or acetamide aldol reactions, which are very difficult to achieve directly with organocatalytic methods.
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
Acetamide, sublimed, 99%