MilliporeSigma
  • Simultaneous determination of aminopyralid, clopyralid, and picloram residues in vegetables and fruits using ultra-performance liquid chromatography/tandem mass spectrometry.
Simultaneous determination of aminopyralid, clopyralid, and picloram residues in vegetables and fruits using ultra-performance liquid chromatography/tandem mass spectrometry.
Journal of AOAC International (2012-06-02)
Yingying Tian, Xingang Liu, Fengshou Dong, Jun Xu, Caihong Lu, Zhiqiang Kong, Yunhao Wang, Yongquan Zheng
ABSTRACT
A sensitive and effective method for the simultaneous quantitative determination of aminopyralid, clopyralid, and picloram in vegetables (eggplant, cucumber, and tomato) and fruits (apple and grape) was developed and validated using ultra-performance LC coupled with MS/MS. The three herbicides were successfully separated and independently confirmed in a single run. Different extraction and cleanup methods were used to optimize the pretreatment processes of the residue analysis method. The final method is straightforward and involves extraction with 1% formic acid-acetonitrile, and no complicated cleanup process is needed. Determination of the compounds was achieved within 3.0 min. Respective average recoveries using this method at four concentration levels (0.05, 0.1, 0.5, and 1.0 mg/kg) ranged from 66.5 to 109.4%, with RSDs in the range of 1.1-19.7% (n = 5) for all analytes. The LODs were below 0.010 mg/kg, and the LOQs did not exceed 0.036 mg/kg, which were lower than the maximum residue limits (MRLs) of 0.5-5.0 mg/kg clopyralid in vegetables and fruits samples, as established by the European Union. This study provides a theoretical basis for China to develop MRLs and an analytical method for aminopyralid, clopyralid, and picloram in vegetables and fruits.
Sigma-Aldrich
Picloram, BioReagent, suitable for plant cell culture
Supelco
Picloram, PESTANAL®, analytical standard
Supelco
Clopyralid, PESTANAL®, analytical standard