MilliporeSigma
  • A luminescent nanoscale metal-organic framework with controllable morphologies for spore detection.
A luminescent nanoscale metal-organic framework with controllable morphologies for spore detection.
Chemical communications (Cambridge, England) (2012-05-26)
Hui Xu, Xingtang Rao, Junkuo Gao, Jiancan Yu, Ziqi Wang, Zhongshang Dou, Yuanjing Cui, Yu Yang, Banglin Chen, Guodong Qian
ABSTRACT
A nanoscale MOF material NMOF 1 with controllable morphologies is realized whose morphology control has been simulated based on the BFDH method. The targeted NMOF 1 exhibits highly sensitive, selective and instant "turn-on" sensing of bacterial endospores.
Product Description
Supelco
2,6-Pyridinedicarboxylic acid concentrate, 0.02 M C7H5NO4 in water (0.04N), suitable for ion chromatography, eluent concentrate
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%