MilliporeSigma
  • Divergent effects of a CLA-enriched beef diet on metabolic health in ApoE-/- and ob/ob mice.
Divergent effects of a CLA-enriched beef diet on metabolic health in ApoE-/- and ob/ob mice.
The Journal of nutritional biochemistry (2012-05-26)
Clare M Reynolds, Sinead Toomey, Rachael McBride, Jolene McMonagle, Melissa J Morine, Orina Belton, Aidan P Moloney, Helen M Roche
ABSTRACT
Conjugated linoleic acid (CLA) is found naturally in meat and dairy products, and represents a potential therapeutic functional nutrient. However, given the discrepancies in isomer composition and concentration, controversy surrounds its proposed antidiabetic, antiobesity effects. This study focused on the effects of CLA-enriched beef (composed predominantly of c9, t11-CLA) in two separate models of metabolic disease: proatherosclerotic ApoE(-/-) mice and diabetic, leptin-deficient ob/ob mice. Animals were fed CLA-enriched beef for 28 days, and markers of the metabolic syndrome and atherosclerosis were assessed. Comprehensive hepatic transcriptomic analysis was completed to understand divergent metabolic effects of CLA. CLA-enriched beef significantly reduced plasma glucose, insulin, nonesterified fatty acid and triacylglycerol and increased adiponectin levels in ob/ob mice. In contrast, plasma lipid profiles and glucose homeostasis deteriorated and promoted atherosclerosis following the CLA-enriched beef diet in ApoE(-/-) mice. Hepatic transcriptomic profiling revealed divergent effects of CLA-enriched beef on insulin signaling and lipogenic pathways, which were adversely affected in ApoE(-/-) mice. This study demonstrated clear divergence in the effects of CLA. CLA-enriched beef improved metabolic flexibility in ob/ob mice, resulting in enhanced insulin sensitivity. However, CLA-enriched diet increased expression of lipogenic genes, resulting in inefficient fatty acid storage which increases lipotoxicity in peripheral organs, and led to profound metabolic dysfunction in ApoE(-/-) mice. While CLA may have potential health effects, in some circumstances, caution must be exercised in presenting this bioactive lipid as a potential functional food for the treatment of metabolic disease.
Product Description
Supelco
Conjugated (9Z,11E)-Linoleic acid, analytical standard
Supelco
Conjugated (9E,11E)-Linoleic acid, analytical standard