MilliporeSigma
Multidrug-resistant tuberculosis that required 2 years for diagnosis.
BMJ case reports (2012-05-19)
Shuichi Yano, Kanako Kobayashi, Toshikazu Ikeda
ABSTRACT
Isoniazid (H) or rifampicin (R) mono-resistant disease can be treated easily and effectively with first-line drugs, while combined H and R resistance (ie, multidrug-resistant tuberculosis (MDRTB)) requires treatment with at least four agents, including a quinolone and an injectable agent. Drug-resistant Mycobacterium tuberculosis strains are reported to be extremely difficult to cultivate invitro. The authors report a case of MDRTB that required 2 years for diagnosis, and was detected only in sputum culture on solid medium. Physicians should consider MDRTB if TB is suspected but pathogens are not detected.
Sigma-Aldrich
4-Aminosalicylic acid, 99%
Sigma-Aldrich
Sodium 4-aminosalicylate dihydrate, ≥97.0% (T)