MilliporeSigma
  • p-Cresol: a sex pheromone component identified from the estrous urine of mares.
p-Cresol: a sex pheromone component identified from the estrous urine of mares.
Journal of chemical ecology (2012-05-18)
Vincas Būda, Raimondas Mozūraitis, Jonas Kutra, Anna-Karin Borg-Karlson
ABSTRACT
Previously it was shown that m- and p-cresols in the urine of mares exhibits a temporally reproducible pattern that is dependent on ovarian activity and, thus, provides information about the timing of ovulation. New behavioral data demonstrate 1) that stallions spend significantly more time sniffing p-cresol as compared to o-, and m-cresols, and, 2) that the extent of stallions' erections differ significantly in response to different types of samples. The lowest erection level was recorded for the pure-water control, a moderate erection level was elicited by the urine of diestrous mares, and the highest erection level was elicited by urine of a diestrous mare containing synthetic p-cresol at a quantity equivalent to half of the amount of p-cresol found in the urine of estrous mares. Consequently, p-cresol is at least one of the components of a horse sex pheromone.
Product Description
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
p-Cresol, ≥99%, FG
Supelco
4-Methylphenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
p-Cresol, 99%