MilliporeSigma
  • New approach to the preparation of bicyclo octane derivatives via the enantioselective cascade reaction catalyzed by chiral diamine-Ni(OAc)2 complex.
New approach to the preparation of bicyclo octane derivatives via the enantioselective cascade reaction catalyzed by chiral diamine-Ni(OAc)2 complex.
Organic & biomolecular chemistry (2012-05-17)
Wenyi Li, Xiaodong Liu, Zhifeng Mao, Qiao Chen, Rui Wang
ABSTRACT
A highly efficient catalyst system assembled from enantiomerically pure diaminocyclohexane and Ni(OAc)(2) is, for the first time, used to catalyze the cascade Michael-Henry reaction of various diones and substituted nitroalkenes. A series of polyfunctionalized bicyclo[3.2.1]octane derivatives containing four stereogenic centers are prepared with excellent enantioselectivities (up to >99% ee) and diastereoselectivities (up to 50 : 1 dr) with high yields. In addition, via this chiral diamine-Ni(OAc)(2) catalyst system, the base-induced epimerization leading to the decrease of stereoselectivity can be prevented.
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Octane, reagent grade, 98%