MilliporeSigma
  • Concentration- and time-dependent eccentric changes in circular dichroism of saccharide-linked ethynylpyridine oligomer with copper(II) ions.
Concentration- and time-dependent eccentric changes in circular dichroism of saccharide-linked ethynylpyridine oligomer with copper(II) ions.
The Journal of organic chemistry (2012-05-15)
Hajime Abe, Yuki Ohishi, Masahiko Inouye
ABSTRACT
We studied the preparation and circular dichroism (CD) behavior of 2,6-pyridylene ethynylene octameric oligomer linked to a β-d-glucoside moiety with a C20-alkylene chain. The addition of Cu(OTf)(2) salt led to a remarkable enhancement of the first CD band of the oligomer, and an unexpected concentration and time dependency were observed. For example, when 1.0 × 10(-3) M of Cu(OTf)(2) was added to a 1,2-dichloroethane solution of the oligomer (5.0 × 10(-4) M, unit concentration), the CD band appeared in the positive at first and gradually inverted into the negative with time.
Product Description
Supelco
1,2–Dichloroethane solution, certified reference material, 200 μg/mL in methanol
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%