MilliporeSigma
  • Distinct phytochrome actions in nonvascular plants revealed by targeted inactivation of phytobilin biosynthesis.
Distinct phytochrome actions in nonvascular plants revealed by targeted inactivation of phytobilin biosynthesis.
Proceedings of the National Academy of Sciences of the United States of America (2012-05-09)
Yu-Rong Chen, Yi-shin Su, Shih-Long Tu
ABSTRACT
The red/far-red light photoreceptor phytochrome mediates photomorphological responses in plants. For light sensing and signaling, phytochromes need to associate with open-chain tetrapyrrole molecules as the chromophore. Biosynthesis of tetrapyrrole chromophores requires members of ferredoxin-dependent bilin reductases (FDBRs). It was shown that LONG HYPOCOTYL 2 (HY2) is the only FDBR in flowering plants producing the phytochromobilin (PΦB) for phytochromes. However, in the moss Physcomitrella patens, we found a second FDBR that catalyzes the formation of phycourobilin (PUB), a tetrapyrrole pigment usually found as the protein-bound form in cyanobacteria and red algae. Thus, we named the enzyme PUB synthase (PUBS). Severe photomorphogenic phenotypes, including the defect of phytochrome-mediated phototropism, were observed in Physcomitrella patens when both HY2 and PUBS were disrupted by gene targeting. This indicates HY2 and PUBS function redundantly in phytochrome-mediated responses of nonvascular plants. Our studies also show that functional PUBS orthologs are found in selected lycopod and chlorophyte genomes. Using mRNA sequencing for transcriptome profiling, we demonstrate that expression of the majority of red-light-responsive genes are misregulated in the pubs hy2 double mutant. These studies showed that moss phytochromes rapidly repress expression of genes involved in cell wall organization, transcription, hormone responses, and protein phosphorylation but activate genes involved in photosynthesis and stress signaling during deetiolation. We propose that, in nonvascular plants, HY2 and PUBS produce structurally different but functionally similar chromophore precursors for phytochromes. Holophytochromes regulate biological processes through light signaling to efficiently reprogram gene expression for vegetative growth in the light.
Sigma-Aldrich
R-Phycoerythrin, BioReagent, passes test for gel electrophoresis