MilliporeSigma
  • Identification of factors regulating Escherichia coli 2,3-butanediol production by continuous culture and metabolic flux analysis.
Identification of factors regulating Escherichia coli 2,3-butanediol production by continuous culture and metabolic flux analysis.
Journal of microbiology and biotechnology (2012-05-09)
Mingshou Lu, Soojin Lee, Borim Kim, Changhun Park, Minkyu Oh, Kyungmoon Park, Sang Yup Lee, Jinwon Lee
ABSTRACT
2,3-Butanediol (2,3-BDO) is an organic compound with a wide range of industrial applications. Although Escherichia coli is often used for the production of organic compounds, the wild-type E. coli does not contain two essential genes in the 2,3-BDO biosynthesis pathway, and cannot ferment 2,3-BDO. Therefore, a 2,3-BDO biosynthesis mutant strain of Escherichia coli was constructed and cultured. To determine the optimum culture factors for 2,3-BDO production, experiments were conducted under different culture environments ranging from strongly acidic to neutral pH. The extracellular metabolite profiles were obtained using high-performance liquid chromatography (HPLC), and the intracellular metabolite profiles were analyzed by ultra-performance liquid chromatography and quadruple time-of-flight mass spectrometry (UPLC/ Q-TOF-MS). Metabolic flux analysis (MFA) was used to integrate these profiles. The metabolite profiles showed that 2,3-BDO production favors an acidic environment (pH 5), whereas cell mass favors a neutral environment. Furthermore, when the pH of the culture fell below 5, both the cell growth and 2,3-BDO production were inhibited.
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
(2S,3S)-(+)-2,3-Butanediol, 97%
Sigma-Aldrich
(2R,3R)-(−)-2,3-Butanediol, 97%
Sigma-Aldrich
meso-2,3-Butanediol, 99%