MilliporeSigma
  • In vitro activity of mastoparan-AF alone and in combination with clinically used antibiotics against multiple-antibiotic-resistant Escherichia coli isolates from animals.
In vitro activity of mastoparan-AF alone and in combination with clinically used antibiotics against multiple-antibiotic-resistant Escherichia coli isolates from animals.
Peptides (2012-05-09)
Chun-Hsien Lin, Roger F Hou, Ching-Lin Shyu, Wei-Yau Shia, Chuen-Fu Lin, Wu-Chun Tu
ABSTRACT
The in vitro activity of mastoparan-AF, an amphipathic antimicrobial peptide isolated from the hornet venom of Vespa affinis, alone and in combination with various clinically used antibiotics, was investigated against 21 Escherichia coli isolates/strains. Most E. coli isolates tested were detected containing multiple-antimicrobial resistance genes. Antimicrobial activity of mastoparan-AF was measured by MIC, MBC, time-kill kinetic assay and chequerboard titration method. Mastoparan-AF exhibited potent antimicrobial activity against most multiple-antibiotic-resistant E. coli isolates at the concentrations ranging from 4 to 16 μg/ml. Combination studies showed that mastoparan-AF acts synergistically with certain antibiotics, i.e., cephalothin or gentamicin, against some multiple-antibiotic-resistant E. coli isolates. In conclusion, mastoparan-AF alone or in combination with other antibiotics could be promising as alternatives for combating multiple-antibiotic-resistant E. coli in future clinical applications.
Sigma-Aldrich
Cephalothin sodium salt