MilliporeSigma
  • Effect of chronic exposure to acetaminophen and lincomycin on Japanese medaka (Oryzias latipes) and freshwater cladocerans Daphnia magna and Moina macrocopa, and potential mechanisms of endocrine disruption.
Effect of chronic exposure to acetaminophen and lincomycin on Japanese medaka (Oryzias latipes) and freshwater cladocerans Daphnia magna and Moina macrocopa, and potential mechanisms of endocrine disruption.
Chemosphere (2012-05-09)
PanGyi Kim, Yena Park, Kyunghee Ji, Jihyun Seo, Sangwoo Lee, Kyunghee Choi, Younglim Kho, Jeongim Park, Kyungho Choi
ABSTRACT
Chronic toxicity of acetaminophen and lincomycin were evaluated using freshwater organisms including two crustaceans (Daphnia magna and Moina macrocopa) and a fish (Oryzias latipes). H295R, a human adrenal cell was also used to understand the effects on steroidogenesis. In 21 d D. magna exposure, survival NOEC was found at 5.72 mg L(-1) and no reproduction related effects were noted at this level of exposure to acetaminophen, while 21 d survival or growth effects were not observed even at the highest exposure levels (153 mg L(-1)) for lincomycin. In the chronic fish toxicity test, significant reduction in juvenile survival was observed at 30 d post-hatch (dph) at 95 mg L(-1) of acetaminophen, and 0.42 mg L(-1) of lincomycin. After the exposure to both pharmaceuticals, vitellogenin levels tended to increase in male fish at 90 dph. In the eggs which were prenatally exposed to 9.5 mg L(-1) of acetaminophen, reduced hatchability was observed. The results of H295R cell assay showed that both pharmaceuticals could alter steroidogenic pathway and increase estrogenicity. Endocrine disruption potentials and their ecological implication may deserve further studies. Our observations suggest however that ecological risks of both pharmaceuticals are negligible at the concentrations currently found in the environment.
Sigma-Aldrich
Lincomycin hydrochloride, 96.0-102.0% (HPLC)
Supelco
Lincomycin hydrochloride monohydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Lincomycin hydrochloride, ≥90% (TLC)
Sigma-Aldrich
Lincomycin hydrochloride, BioReagent, suitable for cell culture
Lincomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard