MilliporeSigma
  • New host material for high-performance blue phosphorescent organic electroluminescent devices.
New host material for high-performance blue phosphorescent organic electroluminescent devices.
Advanced materials (Deerfield Beach, Fla.) (2012-05-03)
Jong-Kwan Bin, Nam-Sung Cho, Jong-In Hong
ABSTRACT
A new carbazole derivative with two carbazole moieties on the C3 and C6 positions of carbazole and triphenylsilane directly linked to the N of carbazole is successfully used as a highly efficient blue phosphorescent host in an organic light-emitting diode (PhOLED). The blue PhOLED, with this new host, gives an extremely high external quantum efficiency.
Sigma-Aldrich
Carbazole, ≥95% (GC)
Supelco
Carbazole, VETRANAL®, analytical standard