MilliporeSigma
  • Primary hyperoxaluria Type 1: indications for screening and guidance for diagnosis and treatment.
Primary hyperoxaluria Type 1: indications for screening and guidance for diagnosis and treatment.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2012-05-02)
Pierre Cochat, Sally-Anne Hulton, Cécile Acquaviva, Christopher J Danpure, Michel Daudon, Mario De Marchi, Sonia Fargue, Jaap Groothoff, Jérôme Harambat, Bernd Hoppe, Neville V Jamieson, Markus J Kemper, Giorgia Mandrile, Martino Marangella, Stefano Picca, Gill Rumsby, Eduardo Salido, Michael Straub, Christiaan S van Woerden
ABSTRACT
Primary hyperoxaluria Type 1 is a rare autosomal recessive inborn error of glyoxylate metabolism, caused by a deficiency of the liver-specific enzyme alanine:glyoxylate aminotransferase. The disorder results in overproduction and excessive urinary excretion of oxalate, causing recurrent urolithiasis and nephrocalcinosis. As glomerular filtration rate declines due to progressive renal involvement, oxalate accumulates leading to systemic oxalosis. The diagnosis is based on clinical and sonographic findings, urine oxalate assessment, enzymology and/or DNA analysis. Early initiation of conservative treatment (high fluid intake, pyridoxine, inhibitors of calcium oxalate crystallization) aims at maintaining renal function. In chronic kidney disease Stages 4 and 5, the best outcomes to date were achieved with combined liver-kidney transplantation.
Sigma-Aldrich
Potassium citrate tribasic monohydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets USP testing specifications
Sigma-Aldrich
Potassium citrate tribasic monohydrate, ≥98% (GC/titration)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, suitable for cell culture
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)