MilliporeSigma
  • Carbon nanohorns as a high-performance carrier for MnO2 anode in lithium-ion batteries.
Carbon nanohorns as a high-performance carrier for MnO2 anode in lithium-ion batteries.
ACS applied materials & interfaces (2012-05-02)
Heng Lai, Jiaxin Li, Zhigao Chen, Zhigao Huang
ABSTRACT
MnO(2) nanoflakes coated on carbon nanohorns (CNHs) has been synthesized via a facile solution method and evaluated as anode for lithium-ion batteries. By using CNHs as buffer carrier, MnO(2)/CNH composite displays an excellent capacity of 565 mA h/g measured at a high current density of 450 mA/g after 60 cylces.
Sigma-Aldrich
Manganese(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Sigma-Aldrich
Manganese(IV) oxide, ReagentPlus®, ≥99%