MilliporeSigma
Live cell imaging of a fluorescent gentamicin conjugate.
Natural product communications (2012-05-02)
Jorge O Escobedo, Yu-Hsuan Chu, Qi Wang, Peter S Steyger, Robert M Strongin
ABSTRACT
Understanding cellular mechanisms of ototoxic and nephrotoxic drug uptake, intracellular distribution, and molecular trafficking across cellular barrier systems aids the study of potential uptake blockers that preserve sensory and renal function during critical life-saving therapy. Herein we report the design, synthesis characterization and evaluation of a fluorescent conjugate of the aminoglycoside antibiotic gentamicin. Live cell imaging results show the potential utility of this new material. Related gentamicin conjugates studied to date quench in live kindney cells, and have been largely restricted to use in fixed (delipidated) cells.