MilliporeSigma
  • A single-cell assay of beta-galactosidase activity in Saccharomyces cerevisiae.
A single-cell assay of beta-galactosidase activity in Saccharomyces cerevisiae.
Cytometry (1988-07-01)
K D Wittrup, J E Bailey
ABSTRACT
A novel assay of single-cell exogenous beta-galactosidase activity in Saccharomyces cerevisiae has been developed. Intracellular fluorescence due to the hydrolysis of resorufin-beta-D-galactopyranoside attains a steady state between production of resorufin and its subsequent leakage from the cell. The cells are permeabilized with Triton X-100, and the assay is performed at 0 degrees C. These conditions were chosen to minimize intercellular fluorescence communication. Free resorufin in the extracellular space is bound by bovine serum albumin to prevent its uptake by cells. Two regimes of fluorescence accumulation are observed, one limited by the rate of diffusion of substrate into the cell, and one limited by the rate of enzymatic cleavage of the substrate. A quantitative correlation between fluorescence and beta-galactosidase activity is obtained under optimized assay conditions.
Sigma-Aldrich
Resorufin β-D-galactopyranoside, ~95%