MilliporeSigma
  • Enteric methane emissions and lactational performance of Holstein cows fed different concentrations of coconut oil.
Enteric methane emissions and lactational performance of Holstein cows fed different concentrations of coconut oil.
Journal of dairy science (2012-05-01)
M Hollmann, W J Powers, A C Fogiel, J S Liesman, N M Bello, D K Beede
ABSTRACT
To determine if dietary medium-chain fatty acids (FA; C(8) to C(14)) may mitigate enteric methane emissions, 24 cows were blocked by body size (n=2) and randomly assigned to 1 sequence of dietary treatments. Diets were fed for 35 d each in 2 consecutive periods. Diets differed in concentrations of coconut oil (CNO; ~75% medium-chain FA): 0.0 (control) or 1.3, 2.7, or 3.3% CNO, dry matter basis. The control diet contained 50% forage (74% from corn silage), 16.5% crude protein (60% from rumen-degradable protein), 34% neutral detergent fiber (NDF; 71% from forage), and 28% starch, dry matter basis. Data and sample collections were from d 29 to 35 in environmentally controlled rooms to measure methane (CH(4)) production. Methane emitted was computed from the difference in concentrations of inlet and outlet air and flux as measured 8 times per day. Control cows emitted 464 g of CH(4)/d, consumed 22.9 kg of DM/d, and produced 34.8 kg of solids-corrected milk/d and 1.3 kg of milk fat/d. Treatment with 1.3, 2.7, or 3.3% dietary CNO reduced CH(4) (449, 291, and 253 g/d, respectively), but concomitantly depressed dry matter intake (21.4, 17.9, and 16.2 kg/d, respectively), solids-corrected milk yield (36.3, 28.4, and 26.8 kg/d, respectively), and milk fat yield (1.4, 0.9, and 0.9 kg/d, respectively). The amount of NDF digested in the total tract decreased with increased dietary CNO concentrations; thus, CH(4) emitted per unit of NDF digested rose from 118 to 128, 153, and 166 g/kg across CNO treatments. Dietary CNO did not significantly affect apparent digestibility of CP but increased apparent starch digestibility from 92 to 95%. No FA C(10) or shorter were detected in feces, and apparent digestibility decreased with increasing FA chain length. Coconut oil concentrations of 2.7 or 3.3% decreased yields of milk FA <C(12) and >C(14). The highest milk fat concentration (3.69%; 1.3% CNO) was due to the greatest yields of C(12) to C(16) milk FA. Milk FA concentrations of C(18:2 trans-10,cis-12) were related to increased dietary CNO concentrations and presumably to depressed ruminal NDF digestion. Moderate dietary CNO concentrations (e.g., 1.3%) may benefit lactational performance; however, CNO concentrations greater than or equal to 2.7% depressed dry matter intake, milk yield, milk fat yield, and NDF utilization. If mitigation of enteric CH(4) emissions is due to decreased digestion of dietary NDF, then this will lessen a major advantage of ruminants compared with nonruminants in food-production systems. Thus, CNO has limited use for enteric CH(4) mitigation in lactating dairy cows.
Sigma-Aldrich
Coconut oil from Cocos nucifera, low-melting solid