MilliporeSigma
Frequency-dependent Escherichia coli chemotaxis behavior.
Physical review letters (2012-05-01)
Xuejun Zhu, Guangwei Si, Nianpei Deng, Qi Ouyang, Tailin Wu, Zhuoran He, Lili Jiang, Chunxiong Luo, Yuhai Tu
ABSTRACT
We study Escherichia coli chemotaxis behavior in environments with spatially and temporally varying attractant sources by developing a unique microfluidic system. Our measurements reveal a frequency-dependent chemotaxis behavior. At low frequency, the E. coli population oscillates in synchrony with the attractant. In contrast, in fast-changing environments, the population response becomes smaller and out of phase with the attractant waveform. These observations are inconsistent with the well-known Keller-Segel chemotaxis equation. A new continuum model is proposed to describe the population level behavior of E. coli chemotaxis based on the underlying pathway dynamics. With the inclusion of a finite adaptation time and an attractant consumption rate, our model successfully explains the microfluidic experiments at different stimulus frequencies.