MilliporeSigma
  • Formation mechanism of a new carbamazepine/malonic acid cocrystal polymorph.
Formation mechanism of a new carbamazepine/malonic acid cocrystal polymorph.
International journal of pharmaceutics (2012-04-28)
Waree Limwikrant, Aiko Nagai, Yumi Hagiwara, Kenjirou Higashi, Keiji Yamamoto, Kunikazu Moribe
ABSTRACT
A new 2/1 carbamazepine (CBZ)/malonic acid (MA) cocrystal polymorph form C was formed using a vibrational rod mill, whereas the known cocrystal polymorph form A was prepared using a ball mill. IR measurements showed that the interaction between CBZ and MA in cocrystal form C was formed by amide-carboxylic acid heterosynthons, similar to that in cocrystal form A. However, NMR results showed that the molecular states of CBZ at the dibenzazepine ring appeared to be different, which could be due to variation in either the conjugation of the aromatic rings or the π-π interaction of CBZ. Factors affecting the formation of cocrystal polymorphs, such as heat and force, were investigated to clarify the formation mechanism.
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Supelco
Malonic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium malonate dibasic, ≥97.0% (NT)