MilliporeSigma
  • Antimicrobial effect of linalool and α-terpineol against periodontopathic and cariogenic bacteria.
Antimicrobial effect of linalool and α-terpineol against periodontopathic and cariogenic bacteria.
Anaerobe (2012-04-28)
Soon-Nang Park, Yun Kyong Lim, Marcelo Oliveira Freire, Eugene Cho, Dongchun Jin, Joong-Ki Kook
ABSTRACT
Linalool and α-terpineol exhibited strong antimicrobial activity against periodontopathic and cariogenic bacteria. However, their concentration should be kept below 0.4 mg/ml if they are to be used as components of toothpaste or gargling solution. Moreover, other compounds with antimicrobial activity against periodontopathic and cariogenic bacteria should be used in combination.
Product Description
Supelco
(+)-α-Terpineol, analytical standard
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
α-Terpineol, 90%, technical grade