MilliporeSigma
  • Recurrence of "cured" dural arteriovenous fistulas after Onyx embolization.
Recurrence of "cured" dural arteriovenous fistulas after Onyx embolization.
Neurosurgical focus (2012-04-28)
Peter Adamczyk, Arun Paul Amar, William J Mack, Donald W Larsen
ABSTRACT
Endovascular embolization with Onyx has been increasingly used to treat intracranial and spinal dural arteriovenous fistulas (DAVFs). Several case series have been published in recent years reporting high DAVF cure rates with this technique. Although it is seldom reported, DAVF recurrence may occur despite initial "cure." The authors present 3 separate cases of a recurrent DAVF after successful transarterial Onyx embolization. Despite adequate Onyx penetration into the fistula and draining vein, these cases demonstrate that DAVF recanalization may reappear with filling from previous or newly recruited arterial feeders. Other published reports of DAVF recurrence are examined, and potential contributory factors are discussed. These cases highlight the need for awareness of this possible phenomenon and suggest that follow-up angiography should be considered in patients treated with catheter embolization.
Sigma-Aldrich
Poly(vinyl alcohol-co-ethylene), ethylene 32 mol %