MilliporeSigma
  • Potential of Phragmites australis for the removal of veterinary pharmaceuticals from aquatic media.
Potential of Phragmites australis for the removal of veterinary pharmaceuticals from aquatic media.
Bioresource technology (2012-04-24)
Pedro N Carvalho, M Clara P Basto, C Marisa R Almeida
ABSTRACT
The potential of Phragmites australis was evaluated for the removal of three veterinary drugs, enrofloxacin (ENR), ceftiofur (CEF) and tetracycline (TET), from aquatic mediums. Results showed that the plant promoted the removal of 94% and 75% of ENR and TET, respectively, from wastewater. Microbial abundance estimation revealed that microorganisms were not a major participant. Occurrence of drugs adsorption to plant roots was observed in small extension. Therefore, main mechanisms occurring were drug removal by plant uptake and/or degradation. Present results demonstrated the potential of P. australis-planted beds to be used for removal of pharmaceuticals from livestock and slaughterhouse industries wastewater.
Product Description
Supelco
Ceftiofur hydrochloride, VETRANAL®, analytical standard