MilliporeSigma
  • Investigation of structure-property relationships of polyisobutylene-based biomaterials: Morphology, thermal, quasi-static tensile and long-term dynamic fatigue behavior.
Investigation of structure-property relationships of polyisobutylene-based biomaterials: Morphology, thermal, quasi-static tensile and long-term dynamic fatigue behavior.
Journal of the mechanical behavior of biomedical materials (2012-04-24)
C Götz, G T Lim, J E Puskas, V Altstädt
ABSTRACT
This study examines the morphology, thermal, quasi-static and long-term dynamic creep properties of one linear and three arborescent polyisobutylene-based block copolymers (L_SIBS31, D_IBS16, D_IBS27 and D_IBS33). Silicone rubber, a common biopolymer, was considered as a benchmark material for comparison. A unique hysteretic testing methodology of Stepwise Increasing Load Test (SILT) and Single Load Test (SLT) was used in this study to evaluate the long-term dynamic fatigue performance of these materials. Our experimental findings revealed that the molecular weight of polyisobutylene (PIB) and polystyrene (PS) arms [M(n)(PIB(arm)) and M(n)(PS(arm))], respectively had a profound influence on the nano-scaled phase separation, quasi-static tensile, thermal transition, and dynamic creep resistance behaviors of these PIB-based block copolymers. However, silicone rubber outperformed the PIB-based block copolymers in terms of dynamic creep properties due to its chemically crosslinked structure. This indicates a need for a material strategy to improve the dynamic fatigue and creep of this class of biopolymers to be considered as alternative to silicone rubber for biomedical devices.
Sigma-Aldrich
Polyisobutylene, average Mw ~1,000,000, average Mn ~600,000 by GPC/MALLS, average Mv ~1,200,000
Sigma-Aldrich
Polyisobutylene, average Mw ~500,000, average Mn ~200,000 by GPC/MALLS, average Mv ~420,000