MilliporeSigma
  • [Analysis on the compositions of volatile oil of Alpinia henryi with GC-MS method].
[Analysis on the compositions of volatile oil of Alpinia henryi with GC-MS method].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-04-17)
Hua-Zhen Qin, Lei Liu, Xiao-Qian Wang, Ying Liu, Teng-Fei Yu
ABSTRACT
To study the components of volatile oil of Alpinia henryi. The volatile oil was extracted by steam distillation method,used gaseous phase-mass spectrum combination method (GC-MS) to analyze the components of volatile oil. 58 kinds of components were isolated, among them 42 were identified and determined the relative content. This study provides a basis for the development and utilization of Alpinia henryi.
Sigma-Aldrich
(+)-Fenchone, purum, ≥98.0% (sum of enantiomers, GC)
Supelco
(+)-Fenchone, analytical standard
Sigma-Aldrich
L-Fenchone, ≥98%
Sigma-Aldrich
(1R)-(−)-Fenchone, ≥98%