MilliporeSigma
  • p-Sulfonated calix[6]arene modified graphene as a 'turn on' fluorescent probe for L-carnitine in living cells.
p-Sulfonated calix[6]arene modified graphene as a 'turn on' fluorescent probe for L-carnitine in living cells.
Chemical communications (Cambridge, England) (2012-04-14)
Xiaowei Mao, Demei Tian, Haibing Li
ABSTRACT
p-Sulfonated calix[6]arene modified graphene has been firstly synthesized and provided as a 'turn on' probe for L-carnitine through monitoring the fluorescence signal both in vitro and in living cells.
Sigma-Aldrich
Gram′s safranin solution, for microscopy
Sigma-Aldrich
Safranin O, for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0)
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Safranin O, certified by the Biological Stain Commission