MilliporeSigma
  • Animal-level factors affecting ovarian function in Bos indicus heifers treated to synchronize ovulation with intravaginal progesterone-releasing devices and oestradiol benzoate.
Animal-level factors affecting ovarian function in Bos indicus heifers treated to synchronize ovulation with intravaginal progesterone-releasing devices and oestradiol benzoate.
Reproduction in domestic animals = Zuchthygiene (2012-04-14)
S A A Butler, N J Phillips, G B Boe-Hansen, G A Bo, B M Burns, K Dawson, M R McGowan
ABSTRACT
The primary objective of this study was to investigate the impact of animal-level factors including energy balance and environmental/management stress, on the ovarian function of Bos indicus heifers treated to synchronize ovulation. Two-year-old Brahman (BN) (n = 30) and BN-cross (n = 34) heifers were randomly allocated to three intravaginal progesterone-releasing device (IPRD) treatment groups: (i) standard-dose IPRD [Cue-Mate(®) (CM) 1.56 g; n = 17]; (ii) half-dose IPRD [0.78 g progesterone (P(4)); CM 0.78 g; n = 15]; (iii) half-dose IPRD + 300 IU equine chorionic gonadotrophin at IPRD removal (CM 0.78 g + G; n = 14); (iv) and a control group, 2× PGF(2α) [500 μg prostaglandin F(2α) (PGF(2α))] on Day -16 and -2 (n = 18). Intravaginal progesterone-releasing device-treated heifers received 250 μg PGF(2α) at IPRD insertion (Day -10) and IPRD removal (Day -2) and 1 mg oestradiol benzoate on Day -10 and -1. Heifers were managed in a small feedlot and fed a defined ration. Ovarian function was evaluated by ultrasonography and plasma P(4) throughout the synchronized and return cycles. Energy balance was evaluated using plasma insulin-like growth factor 1 (IGF-I) and glucose concentrations. The impact of environmental stressors was evaluated using plasma cortisol concentration. Heifers that had normal ovarian function had significantly higher IGF-I concentrations at commencement of the experiment (p = 0.008) and significantly higher plasma glucose concentrations at Day -2 (p = 0.040) and Day 4 (p = 0.043), than heifers with abnormal ovarian function. There was no difference between the mean pre-ovulatory cortisol concentrations of heifers that ovulated or did not ovulate. However, heifers that ovulated had higher cortisol concentrations at Day 4 (p = 0.056) and 6 (p = 0.026) after ovulation than heifers that did not ovulate.
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%