MilliporeSigma
  • Heme carrier protein 1 transports heme and is involved in heme-Fe metabolism.
Heme carrier protein 1 transports heme and is involved in heme-Fe metabolism.
American journal of physiology. Cell physiology (2012-04-13)
Solange Le Blanc, Michael D Garrick, Miguel Arredondo
ABSTRACT
Heme-Fe is an important source of dietary iron in humans; however, the mechanism for heme-Fe uptake by enterocytes is poorly understood. Heme carrier protein 1 (HCP1) was originally identified as mediating heme-Fe transport although it later emerged that it was a folate transporter. We asked what happened to heme-Fe and folate uptake and the relative abundance of hcp1 and ho1 mRNA in Caco-2 cells after knockdown by transfection with HCP1-directed short hairpin (sh)RNA. Control Caco-2 cells were cultured in bicameral chambers with 0-80 μM heme-Fe for selected times. Intracellular Fe and heme concentration increased in Caco-2 cells reflecting higher external heme-Fe concentrations. Maximum Fe, heme, and heme oxygenase 1 (HO1) expression and activity were observed between 12 and 24 h of incubation. Quantitative RT-PCR for hcp1 revealed that its mRNA decreased at 20 μM heme-Fe while ho1 mRNA and activity increased. When shRNA knocked down hcp1 mRNA, heme-(55)Fe uptake and [(3)H]folate transport mirrored the mRNA decrease, ho1 mRNA increased, and flvcr mRNA was unchanged. These data argue that HCP1 is involved in low-affinity heme-Fe uptake not just in folate transport.
Sigma-Aldrich
Iron(III) citrate, technical grade
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture
Supelco
Iron(III) citrate tribasic monohydrate, 18-20% Fe basis (T)