MilliporeSigma
  • Thiotepa and melphalan based single, tandem, and triple high dose therapy and autologous stem cell transplantation for high risk neuroblastoma.
Thiotepa and melphalan based single, tandem, and triple high dose therapy and autologous stem cell transplantation for high risk neuroblastoma.
Pediatric blood & cancer (2012-04-12)
Ulla M Saarinen-Pihkala, Liisa Hovi, Antti Koivusalo, Kirsi Jahnukainen, Riitta Karikoski, Hannu Sariola, Sakari Wikström
ABSTRACT
Outcome of high risk neuroblastoma (NBL) remains unsatisfactory in spite of intensive treatment efforts. Consolidation with high-dose (HD) chemotherapy and autologous stem cell transplantation (ASCT) has been intensified with tandem and triple cycles with promising results. Our purpose was to improve the outcome with two or three HD-consolidations. Thirty six children with high risk NBL, diagnosed 1995-2010, had intensive induction and surgery, and were stratified to single, tandem or triple HD-therapy and ASCT, followed by local irradiation and cis-retinoic acid. In inoperable patients surgery was facilitated by preoperative HD-melphalan. Long-term outcome of our old cohort from 1987-1994 was updated. Ten year event-free survival (EFS) from diagnosis was 0.44+/-0.10 of the old and 0.43+/-0.085 of the new cohort. EFS from the last ASCT was 0.53 +/-0.12 and 0.48+/-0.091, respectively. Preoperative HD-melphalan rendered 73% of bulky primaries operable in the new cohort. The 5-yr EFS from ASCT was 0.46+/-0.15 for single and 0.73+/-0.15 for tandem ASCT (P = 0.19). All triple ASCT patients, selected by poor/slow response, relapsed or died. Thiotepa- and melphalan based HD regimens, with or without total body irradiation (TBI), appeared to give an outcome comparable to major NBL study groups with acceptable toxicity. Tandem HD therapy gave a 5-year EFS of 73%, whereas a third HD consolidation did not offer any additional advantage for ultra high risk patients with slow response. Pediatr Blood Cancer 2012; 59: 1190-1197. © 2012 Wiley Periodicals, Inc.
Sigma-Aldrich
Thio-TEPA