MilliporeSigma
  • Solid-phase synthesis of asymmetrically branched sequence-defined poly/oligo(amidoamines).
Solid-phase synthesis of asymmetrically branched sequence-defined poly/oligo(amidoamines).
The Journal of organic chemistry (2012-04-11)
Felix Wojcik, Simone Mosca, Laura Hartmann
ABSTRACT
We present for the first time the synthesis of asymmetrically branched sequence-defined poly/oligo(amidoamines) (PAAs) using solid-phase synthesis with the capability of introducing diversity at the side chains. We introduce two new versatile (diethylenetriamine) building blocks for solid-phase synthesis bearing Fmoc/Boc and Fmoc/Alloc protecting groups expanding recently used Fmoc/Boc protecting group strategy for linear PAAs to an Fmoc/Alloc/Boc strategy. This allows for orthogonal on-resin cleavage of Fmoc and Alloc protecting groups during solid-phase synthesis of PAAs with backbones differing in chain length and sequence. With these structures we then demonstrate the potential for generating asymmetrical branching by automated multiple on-resin cleavage of Alloc protecting groups as well as the introduction of side chains varying in length and number. Such systems have high potential as nonviral vectors for gene delivery and will allow for more detailed studies on the correlation between the degree of branching of PAAs and their resulting biological properties.
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%