MilliporeSigma
  • Training on synthetic ethylene-vinyl acetate bench model allows novice medical students to acquire suture skills.
Training on synthetic ethylene-vinyl acetate bench model allows novice medical students to acquire suture skills.
Acta cirurgica brasileira (2012-03-31)
Rafael Denadai, Rogério Saad-Hossne, Marie Oshiiwa, Erika Malheiros Bastos
ABSTRACT
To assess the acquisition of suture skills by training on ethylene-vinyl acetate bench model in novice medical students. Sixteen medical students without previous surgery experience (novices) were randomly divided into two groups. During one hour group A trained sutures on ethylene-vinyl acetate (EVA) bench model with feedback of instructors, while group B (control) received a faculty-directed training based on books and instructional videos. All students underwent a both pre-and post-tests to perform two-and three-dimensional sutures on ox tongue. All recorded performances were evaluated by two blinded evaluators, using the Global Rating Scale. Although both groups have had a better performance (p<0.05) in the post-test when compared with the pre-test, the analysis of post-test showed that group A (EVA) had a better performance (p<0.05) when compared with group B (control). The ethylene vinyl acetate bench model allowed the novice students to acquire suture skills faster when compared to the traditional model of teaching.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor