MilliporeSigma
  • Study of colloidal particle Brownian aggregation by low-coherence fiber optic dynamic light scattering.
Study of colloidal particle Brownian aggregation by low-coherence fiber optic dynamic light scattering.
Journal of colloid and interface science (2012-03-27)
Hui Xia, Ru Yi Pang, Rui Zhang, Cai Xia Miao, Xiao Yun Wu, Xue Shun Hou, Cheng Zhong
ABSTRACT
The aggregation kinetics of particles in dense polystyrene latex suspensions is studied by low-coherence fiber optic dynamic light scattering. Low-coherence fiber optic dynamic light scattering is used to measure the hydrodynamic radius of the aggregates. The aggregation kinetics data obtained can be fitted into a single exponential function, which is the characteristic of slow aggregation. It is found that the aggregation rate of particles increased with higher electrolyte levels and with larger particle concentrations. The experimental results can be explained by use of the Derjaruin-Landau-Verwey-Overbeer (DLVO) theory.
Sigma-Aldrich
Polystyrene, average Mw ~280,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Sigma-Aldrich
Polystyrene, average Mw 35,000
Sigma-Aldrich
Polystyrene, average Mw ~400,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000