MilliporeSigma
  • Hexafluorobenzene in comparison with perfluoro-15-crown-5-ether for repeated monitoring of oxygenation using 19F MRI in a mouse model.
Hexafluorobenzene in comparison with perfluoro-15-crown-5-ether for repeated monitoring of oxygenation using 19F MRI in a mouse model.
Magnetic resonance in medicine (2012-03-24)
Lionel Mignion, Julie Magat, Olivier Schakman, Etienne Marbaix, Bernard Gallez, Bénédicte F Jordan
ABSTRACT
Hexafluorobenzene (HFB) and perfluoro-15-crown-5-ether (15C5) were compared as fluorine reporter probes of tissue oxygenation using (19)F MRI for dynamic assessment of muscle oxygenation, with special focus on muscle tissue toxicity of the probes, and consecutive alteration of animal behavior. The latter were also compared in terms of sensitivity to changes in oxygenation as well as of signal-to-noise ratio for accurate pO(2) measurements. For that purpose, mouse muscles were imaged at 11.7 T, at 2- and 36-h after intramuscular injection of HFB or 15C5. Histological analysis of the muscle tissue revealed a lack of toxicity for 15C5 from 2 up to 36-h postinjection, whereas HFB induced tissue necrosis, blood clots and thrombosis as soon as 24-h postinjection. This muscle toxicity led to a limitation in mice mobility 24-h after injection of HFB as evidenced by behavioral testing (open-field, grip strength, and catwalk tests), which was not the case after 15C5 intramuscular injection. Finally, pO(2) measurements assessed 2-h postinjection showed consistent values with both probes, evidencing cross-validation of the (19)F MRI oximetry technique for acute measurements. However, the measurement at 36-h was hampered for HFB, which showed significant lower values of muscle pO(2), whereas 15C5 was able to reliably assess muscle pO(2) at 36-h postinjection.
Sigma-Aldrich
Hexafluorobenzene, 99%
Sigma-Aldrich
Hexafluorobenzene, ≥99.5%, NMR grade