MilliporeSigma
  • A hard-template method for the preparation of IrO(2) , and its performance in a solid-polymer-electrolyte water electrolyzer.
A hard-template method for the preparation of IrO(2) , and its performance in a solid-polymer-electrolyte water electrolyzer.
ChemSusChem (2012-03-24)
Guangfu Li, Hongmei Yu, Wei Song, Meiling Dou, Yongkun Li, Zhigang Shao, Baolian Yi
ABSTRACT
Morphological control by SBA-15: The performance of catalysts for the oxygen evolution reaction (OER) depends strongly on their structural and morphological properties. An IrO(2) nanomaterial with a morphology suitable for the OER is prepared by using a synthetic scheme involving a zeolite template, and shows enhanced activity and stability compared to IrO(2) fabricated by the traditional Adams-fusion method.
Sigma-Aldrich
Iridium(IV) oxide, 99.9% trace metals basis
Pricing and availability is not currently available.