MilliporeSigma
  • Inadequate efficacy of a new formulation of fosmidomycin-clindamycin combination in Mozambican children less than three years old with uncomplicated Plasmodium falciparum malaria.
Inadequate efficacy of a new formulation of fosmidomycin-clindamycin combination in Mozambican children less than three years old with uncomplicated Plasmodium falciparum malaria.
Antimicrobial agents and chemotherapy (2012-03-21)
Miguel Lanaspa, Cinta Moraleda, Sónia Machevo, Raquel González, Beatriz Serrano, Eusebio Macete, Pau Cisteró, Alfredo Mayor, David Hutchinson, Peter G Kremsner, Pedro Alonso, Clara Menéndez, Quique Bassat
ABSTRACT
The combination of fosmidomycin and clindamycin (F/C) is effective in adults and older children for the treatment of malaria and could be an important alternative to existing artemisinin-based combinations (ACTs) if proven to work in younger children. We conducted an open-label clinical trial to assess the efficacy, safety, and tolerability of F/C for the treatment of uncomplicated P. falciparum malaria in Mozambican children <3 years of age. Aqueous solutions of the drugs were given for 3 days, and the children were followed up for 28 days. The primary outcome was the PCR-corrected adequate clinical and parasitological response at day 28. Secondary outcomes included day 7 and 28 uncorrected cure rates and fever (FCT) and parasite (PCT) clearance times. Fifty-two children were recruited, but only 37 patients were evaluable for the primary outcome. Day 7 cure rates were high (94.6%; 35/37), but the day 28 PCR-corrected cure rate was 45.9% (17/37). The FCT was short (median, 12 h), but the PCT was longer (median, 72 h) than in previous studies. Tolerability was good, and most common adverse events were related to the recurrence of malaria. The poor efficacy observed for the F/C combination may be a consequence of the new formulations used, differential bioavailability in younger children, naturally occurring variations in parasite sensitivity to the drugs, or an insufficient enhancement of their effects by naturally acquired immunity in young children. Additional studies should be conducted to respond to the many uncertainties arising from this trial, which should not discourage further evaluation of this promising combination.
Sigma-Aldrich
Fosmidomycin sodium salt hydrate, ≥95% (NMR)